Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ CHOgro™ High Yield Expression System

Catalog No. MIR6270 Shop All Mirus Bio LLC Products
Change view
Click to view available options
CHOgro™ High Yield Expression System; Format: Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MIR6270 CHOgro™ High Yield Expression System; Format: Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MIR6270 Supplier Mirus Bio™ Supplier No. MIR 6270
Add to Cart
Add to Cart

CHOgro™ High Yield Expression System is an advanced system for transient transfection and protein production in suspension CHO cells.

The CHOgro High Yield Expression System is the most advanced and cost-effective transient transfection system for high-yield protein production in suspension CHO cells. Our second-generation system features the CHOgro Titer Enhancer, which provides rapid, industry-leading protein yields.

  • High Yield - Reach higher antibody titers in seven days - faster than the ExpiCHO Expression System
  • Simple Workflow - Same-day transfection, enhancer addition and temperature shift
  • Worry Free - No commercial license required; animal origin-free

Specifications

Cell Type CHO
Sample Type DNA
Promoter None (Promoterless)
Transfection Technique Lipid-based Transfection, Transient Transfection
Includes CHOgro™ Expression Medium (2 x 1 L), CHOgro™ Complex Formation Solution (100 mL), L-Glutamine (100 mL), Poloxamer 188 Solution (100 mL), TransIT-PRO™ Transfection Reagent (1 mL), CHOgro™ Titer Enhancer (20 mL)
Product Type Expression System
Sterility Sterile
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.