Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ CHOgro™ Transfection-Titer Kit

Catalog No. MIR6225 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Cell Type:
CHO
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Type
MIR6225 CHO
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MIR6225 Supplier Mirus Bio™ Supplier No. MIR 6225
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Advanced system for transient transfection and protein production in suspension CHO cells.

The CHOgro High Yield Expression System is the most advanced and cost-effective transient transfection system for high-yield protein production in suspension CHO cells. Our second-generation system features the CHOgro Titer Enhancer, which provides rapid, industry-leading protein yields.

  • High Yield - Reach higher antibody titers in seven days - faster than the ExpiCHO Expression System
  • Simple Workflow - Same-day transfection, enhancer addition and temperature shift
  • Worry Free - No commercial license required; animal origin-free

Specifications

Cell Type CHO
Sample Type DNA
Promoter None (Promoterless)
Transfection Technique Lipid-based Transfection, Transient Transfection
Includes TransIT-PRO™ Transfection Reagent (1 mL), CHOgro™ Titer Enhancer (20 mL)
Product Type Transfection-Titer Kit
Sterility Sterile
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.