Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholecalciferol, Crystal, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18601649 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-649 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-601-649 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CH1345GM

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

CH134, 67-97-0, C27H44O

Spectrum™ Chemical Cholecalciferol, Crystal, USP is also known as toxiferol and is a form of vitamin D. It is used in vitamin supplements. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 67-97-0
Molecular Formula C27H44O
Molecular Weight (g/mol) 384.65
InChI Key QYSXJUFSXHHAJI-FVUVGDFOSA-N
IUPAC Name (1S,3E)-3-{2-[(1R,3aS,4E,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 5 g
UN Number UN2811
Specific Rotation +105° to +112°
Percent Purity 97 to 103%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.