Learn More
Cayman Chemical -Bornyl Acetate 25g
SDP
Supplier:  Cayman Chemical 2961725 G
Encompass_Preferred
An iridoid glycoside with diverse biological activities increases cell viability and migration of BM-EPCs from 0 1-1000 M inhibits apoptosis and reduces levels of MMP-3 and MMP-13 in chondrocytes inhibits LPS-induced nuclear translocation of NF-KB and reduces COX-2 iNOS NF-a and IL-1B mRNA expression in RAW 264 7 cells reduces colonic MPO activity COX-2 and iNOS mRNA expression and disease severity in a mouse model of DSS-induced ulcerative colitis at 100 and 200 mg/kg increases bone mineral content bone mineral density and improves bone microstructure in ovariectomized mice at 40 and 80 mg/kg reduces acetic acid-induced writhing in mice and carrageenan-induced paw edema in rats at 20 and 30 mg/kg decreases macrophage infiltration and wound healing time and increases blood vessel formation in a rat model of wound healing
Catalog No. 50-299-1150
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.