Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Cholesterol
GREENER_CHOICESDP

Catalog No. 7945100
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-451-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 79-451-00 Supplier Tocris Bioscience™ Supplier No. 7945/100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Component of lipid nanoparticles (LNPs); modulator of lipid bilayer fluidity

Cholesterol is a major sterol in all mammalian organisms; it is an essential component that makes up about 20-25% of the structural lipids of cell membranes. Cholesterol determines the permeability and fluidity characteristics of the cell membranes. Cholesterol is an endogenous ligand for estrogen-related receptor α (ERRα). It is also a precursor in several biosynthetic pathways, including steroid hormones and the active form of vitamin D. Cholesterol is a component of lipid nanoparticles (LNPs), and is important in modulating membrane stability, with the effect being context dependent. When combined with lipids that have low gel-liquid crystalline phase transition temperature (Tm), Cholesterol reduces membrane fluidity and increases bilayer thickness; when combined with high Tm lipids, it increases membrane fluidity and narrows the bilayer thickness.

Specifications

Chemical Name or Material (3β)-Cholest-5-en-3-ol
CAS 57-88-5
Quantity 100 mg
Target Lipid Nanoparticles
Molecular Formula C27H46O
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.