Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Cholesterol Assay Buffer, 25 mL
Description
Cholesterol Assay Buffer for use in cholesterol assay kits.
Specifications
Specifications
| Target Molecule | Cholesterol |
| Kit Contents | Cholesterol Assay Buffer, 25 mL |
| Product Type | Wash Solution |
| Format | Bottle |
| Quantity | 25 mL |
| For Use With (Application) | Cholesterol assay |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?