Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Cholesterol Esterase, from porcine pancreas

Catalog No. AAJ64282EXR
Change view
Click to view available options
Quantity:
100 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64282EXR 100 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64282EXR Supplier Thermo Scientific Chemicals Supplier No. J64282EXR
Add to Cart

missing translation for 'validMainframeMsg'

Cholesterol Esterase is used for enzymatic determination of total cholesterol when coupled with cholesterol oxidize in clinical analysis. Cholesterol esterase (CE) is a reversible enzyme that can hydrolyze or synthesize fatty acid esters of cholesterol and other sterols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 U
Molecular Weight (g/mol) 440
Form Lyophilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.