Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Cholesterol, Lipid, 386.7 Da, >98% Purity, 25 g

Catalog No. 502706227
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6227 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6227 Supplier Abcam Supplier No. AB14250125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity water-permeable lipid for use as an emulsifying agent and in liposome preparations.

  • Molecular Weight: 386.7 Da
  • Purity: >98%
  • Composition: 100% Cholesterol
  • Key Characteristics: Water-permeable lipid; major component of biological membranes; inhibits basal channel activity in the brain
  • Applications: Emulsifying agent; liposome preparations
  • Activity: Active in vivo and in vitro

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at ambient temperature.
Shipping Condition Ambient
Target Molecule Cholesterol
Buffer 100% Cholesterol
Form Powder
Formulation 100% Cholesterol
Quantity 25 g
For Use With (Application) Emulsifying agent, liposome preparations
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.