Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholesterol, NF Grade, MP Biomedicals

Catalog No. MP21013801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21013801 100 g
MP21013803 25 g
MP21013805 500 g
ICN10138001 1 kg
MP210138005 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MP21013801 Supplier MP Biomedicals, Inc Supplier No. 02101380.1
Add to Cart
Add to Cart

  • Cholesterol is suitable for use in liposome preparations
  • Cholesterol is a component used for the preparation and study of artificial model membranes
  • One example is a study of valinomycin induced changes in membrane potentials of red blood cell and phospholipid (phosphatidylcholine from egg yolk plus cholesterol) vesicle suspensions
  • Positively charged cyanine dyes were used that fluorimetrically respond to the change in potential

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-88-5
Molecular Formula C27H46O
Molecular Weight (g/mol) 386.664
MDL Number MFCD00003646
InChI Key HVYWMOMLDIMFJA-DPAQBDIFSA-N
Synonym cholesterol, cholesterin, cholest-5-en-3beta-ol, cholesteryl alcohol, cholestrin, cordulan, dusoline, dusoran, provitamin d, cholesterine
PubChem CID 5997
ChEBI CHEBI:16113
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C

Specifications

Melting Point 147°C to 150°C
Color White
Boiling Point 360°C (literature)
Linear Formula C27H46O
Quantity 100 g
Formula Weight 386.67
Grade NF
Physical Form Powder
Chemical Name or Material CHOLESTEROL
Recommended Storage -20°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.