Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholesteryl Hydrogen Succinate, MP Biomedicals™

Catalog No. ICN10137905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10137905 5 g
ICN10137925 25 g
02-101-37980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10137905 Supplier MP Biomedicals, Inc Supplier No. 0210137905
Add to Cart
Add to Cart

Cholesteryl Hydrogen Succinate is a cholesterol derivative that is used as a detergent.

  • Dicarboxylic acid monoester resulting from the condensation of the cholesterol hydroxy group with a carboxy group of succinic acid.
  • Sometimes used as a replacement for cholesterol in protein crystallography.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1510-21-0
Molecular Formula C31H50O4
Molecular Weight (g/mol) 486.74
MDL Number MFCD00037705
InChI Key WLNARFZDISHUGS-MIXBDBMTSA-N
Synonym cholesteryl hemisuccinate, cholesterol hydrogen succinate, cholesteryl succinate, unii-t3j4ks4201, cholesteryl hydrogen succinate, cholest-5-en-3beta-yl hydrogen succinate, 4-3beta-cholest-5-en-3-yloxy-4-oxobutanoic acid, 4-3s,8s,9s,10r,13r,14s,17r-10,13-dimethyl-17-2r-6-methylheptan-2-yl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-yl oxy-4-oxobutanoic acid, cholesterol hemisuccinate, cholesterylhemisuccinate
PubChem CID 65082
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)OC(=O)CCC(O)=O

Specifications

Quantity 5 g
IUPAC Name 4-[[(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl]oxy]-4-oxobutanoic acid
Physical Form Powder
Chemical Name or Material Cholesteryl hydrogen succinate
EINECSNumber 216-147-0
Recommended Storage 15 to 30°C
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.