Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Choline Assay Kit, Fluorescent, Quantitative, 40 nM Sensitivity, 200 Tests
Description
- Detection method: Fluorescent
- Sensitivity: 40 nM
- Assay type: Quantitative
- Sample types: Tissue lysate, urine, plasma, serum, other biological fluids, and cell lysate
- Compatible instrumentation: Microplate reader
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| Detection Method | Fluorescence |
| No. of Reactions | 200 Tests |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Choline |
| Detection Location | In-Solution Detection |
| Sample Type | Tissue Lysate, Urine, Plasma, Serum, Other biological fluids, Cell Lysate |
| Quantity | 200 Tests |
| For Use With (Application) | Assay Choline levels |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?