Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Bromide 98.0+%, TCI America™
SDP

Catalog No. C032825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C032825G 25 g
C0328500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C032825G Supplier TCI America Supplier No. C032825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1927-06-6
Molecular Formula C5H14BrNO
Molecular Weight (g/mol) 184.08
MDL Number MFCD00054244
InChI Key JJCWKVUUIFLXNZ-UHFFFAOYSA-M
Synonym (2-Hydroxyethyl)trimethylammonium Bromide
PubChem CID 74724
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;bromide
SMILES C[N+](C)(C)CCO.[Br-]

Specifications

Melting Point 298°C
Color White
Quantity 25 g
Formula Weight 184.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Choline Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.