Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Chloride Dietary Supplement, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18601679 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-679 2.5 kg Poly Pail
18-601-678 500 g Poly Bottle
18-601-680 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-601-679 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C20582.5KG
Add to Cart
Add to Cart

C2058, 67-48-1, C5H14ClNO

Spectrum™ Chemical Choline Chloride, USP Dietary Supplement is an essential nutrient and typically grouped with B-complex vitamins. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 67-48-1
Molecular Formula C5H14ClNO
Molecular Weight (g/mol) 139.62
MDL Number MFCD00011721
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
IUPAC Name (2-hydroxyethyl)trimethylazanium chloride
SMILES [Cl-].C[N+](C)(C)CCO

Specifications

CAS Max % 100%
Quantity 2.5 kg
Percent Purity 99 to 100.5%
Grade USP Dietary Supplement
Ignition Residue 0.05%
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.