Learn More
Cellular Engineering Technologies Chondrogenic Differentiation Media (450mL)
Supplier:  Cellular Engineering Technologies MR1008
Encompass_Preferred
Human Chondrogenic Differentiation Media is designed to support the differentiation of adipose or bone marrow-derived mesenchymal stem cells into cartilage-producing cells. Chondrogenic differentiation must be conducted in pellet culture, taking approximately 28 days.Media is shipped with gel packs.
Catalog No. NC2753074