Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chondroitin sulfate, 90+%

Catalog No. AAJ6034106
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6034106 5 g
AAJ6034103 1 g
AAJ6034114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6034106 Supplier Thermo Scientific Chemicals Supplier No. J6034106
Add to Cart

missing translation for 'validMainframeMsg'

Form: Powder

Chondroitin sulfate is used in a wide variety of areas of research such as bioengineering. Chondroitin sulfate may be used as a biomaterial copolymer or surface derivatization reagent in the development of drug delivery vehicles, tissue engineering devices and bioscaffolds. Chondroitin sulfate may be used for the development of biocompatible structures such as hydrogels, sponges, biofilms, microspheres and micelles. Used in the treatment of coronary atherosclerosis, myocardial ischemia, myocardial infraction. Chondroitin sulfate is a sulfated (in different positions) glycosaminoglycan (GAG), composed of repeating and alternating disaccharide units of D-glucuronic acid and N-acetylgalactosamine. Depending on the position of sulfate, Chondroitin sulfate is named as chondroitin sulfate A (sulfate on carbon 4 of the N-acetylgalactosamine (GalNAc) sugar, C (carbon 6 of the GalNAc sugar), D (carbon 2 of the glucuronic acid and 6 of the GalNAc sugar) and E (carbons 4 and 6 of the GalNAc sugar). The CAS # given is the representation of core component of this product.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chondroitin sulfate
Color White
Quantity 5 g
MDL Number MFCD00146419
Merck Index 14,2214
Synonym chondroitin sulfate, chondroitin sulphate, chonsurid, chondroitin sulfate c, chondroitin polysulfate, chondroitin sulfates, chondroitin 4-sulfate, chondroitin 6-sulfate, chondroitin, hydrogen sulfate, chondroitin sulfuric acid
Solubility Information Soluble in water.
InChI Key KXKPYJOVDUMHGS-OSRGNVMNSA-N
SMILES CC(=O)NC1C(C(C(OC1O)OS(=O)(=O)O)O)OC2C(C(C(C(O2)C(=O)O)O)O)O
IUPAC Name (2S,3S,4S,5R,6R)-6-[(2R,3R,4R,5R,6R)-3-acetamido-2,5-dihydroxy-6-sulfooxyoxan-4-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
PubChem CID 24766
Percent Purity ≥90%
Physical Form Powder
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.