Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chondroitin Sulfate Sodium Salt Dietary Supplement, USP, Spectrum™ Chemical
SDP

Catalog No. 18601684 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-684 2.5 kg Poly Pail
18-601-683 500 g Poly Bottle
18-601-685 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-601-684 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C16102.5KG
Add to Cart
Add to Cart
Request a Quote

C1610, 9082-07-9

Spectrum™ Chemical Chondroitin Sulfate Sodium Salt, USP Dietary Supplement is the sodium salt of a sulfated glycosaminoglycan (GAG) composed of a chain of alternating sugars (N-acetylgalactosamine and glucuronic acid) and is used for osteoarthritis. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9082-7-9

Specifications

CAS Max % 100%
Quantity 2.5 kg
Grade USP Dietary Supplement
Ignition Residue 20%-30%
Loss on Drying 12%
Packaging Poly Pail
Specific Rotation -20.0° to -30.0°
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.