Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Chorionic Gonadotropin alpha Chain (hCG alpha) Native Protein
SDP

Catalog No. N19953510U Shop All Novus Products
Change view
Click to view available options
Quantity:
1000 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N19953510U 1000 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N19953510U Supplier R&D Systems™ Supplier No. NBP1995351000UG

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: SDS-Page, Bioactivity

Native bioactive protein containing amino acids for Human hCG Alpha (Intact)

The Chorionic Gonadotropin alpha Chain (hCG alpha) Native Protein is derived from Urine of post-menopausal women. The Chorionic Gonadotropin alpha Chain (hCG alpha) Native Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration Lyoph
For Use With (Application) SDS-PAGE, Bioactivity
Gene ID (Entrez) 1081
Molecular Weight (g/mol) 36.7 kDa
Quantity 1000 μg
Source Urine of post-menopausal women
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol CGA
Specific Reactivity Chorionic Gonadotropin alpha Chain (hCG alpha)
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute in dH2O to a concentration of 0.1 - 1.0 mg/mL. The solution can then be diluted into other aqueous buffers and stored at 4°C for 1 week or -20°C for long-term storage.
Buffer Sterile filtered and lyophilized with no additives
Product Line Novus
Purity or Quality Grade >98%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.