Learn More
CPC Scientific H-Thr-Cys-Asp-Asp-Pro-Arg-Phe-Gln-Asp-Ser-Ser-OH (trifluoroacetate salt) 1MG
Supplier: CPC Scientific HCG001A
This highly immunogenic undecapeptide generates hCGspecific antibodies ONE-LETTER SEQUENCE: TCDDPRFQDSSMOLECULAR FORMULA: C50H76N16O21S1MOLECULAR WEIGHT:1269.32STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [165174-59-4], RESEARCH AREA: ImmunomodulationREFERENCES: K.S.N.Iyer et al., Int. J. Pept. Protein Res., 39, 137 (1992)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.