Learn More
Carbosynth LLC. CHORIONIC GONADOTROPIN HUMAN
SDP
Supplier:  Carbosynth LLC. HOR2502.5MG
Encompass_Preferred
HcG, 2.5mg; Human chorionic gonadotropin (hCG) is a peptide hormone produced in pregnancy, that is made by the embryo soon after conception and later by the syncytiotrophoblast (part of the placenta). Physical Appearance: Sterile Filtered White lyophilized (freeze-dried) powder.Formulation: The hCG was lyophilized with no additives. MFG: PROSPEC BIO
Catalog No. NC1631697
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.