Learn More
Invitrogen™ Chp Monoclonal Antibody (JNC1)

Description
MA1-40274 detects CHIPS from S. aureus samples. MA1-40274 has been successfully used in flow cytometry, ELISA, immunohistochemistry (frozen), immunohistochemistry (paraffin), and Western blot applications. The MA1-40274 immunogen is the N-terminus of CHIPS.
Specifications
Specifications
| Antigen | Chp |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | JNC1 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and no preservative |
| Gene Alias | calcineurin B homolog; calcineurin B homologous protein 1; Calcineurin B-like protein; calcineurin homologous protein; calcineurin like EF-hand protein 1; calcineurin-like EF hand protein 1; calcineurin-like EF-hand protein 1; calcium binding protein P22; calcium-binding protein CHP; Calcium-binding protein p22; CHIPS; CHP; CHP1; EF-hand calcium-binding domain-containing protein p22; p22; p24; Sid470p; SLC9A1 binding protein; SLC9A1BP |
| Host Species | Mouse |
| Immunogen | CHIPS N-Terminus. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.