Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575494
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575494 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575494 Supplier Invitrogen™ Supplier No. PA575494
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

CHP2 (Calcineurin B homologous protein 2), also known as Hepatocellular carcinoma-associated antigen 520, is a 196 amino acid protein that contains four EF-hand domains and plays a potential role in transmembrane Na+/H+ exchange. By binding to and activating NHE-1, CHP2 increases the pH and works to protect cells from serum deprivation-induced death. Though not typically detected in normal tissues, CHP2 is highly expressed in malignantly transformed cells and is therefore considered to be a tumor-associated antigen. Ectopic expression of CHP2 promotes proliferation of HEK293 cells and knockdown of CHP2 mRNA in HepG2 cells inhibits cell proliferation. Like calcineurin B, CHP2 can bind to and stimulate phosphatase activity of calcineurin A and activate the calcineurin/NFAT signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHP2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CHP2
Gene Accession No. O43745, Q810D1, Q9D869
Gene Alias 2010110P09Rik; calcineurin B homologous protein 2; calcineurin like EF-hand protein 2; calcineurin-like EF hand protein 2; calcineurin-like EF-hand protein 2; Cbhp2; CHP2; chp-2; HCA520; hepatocellular carcinoma antigen gene 520; hepatocellular carcinoma-associated antigen 520; Hepatocellular carcinoma-associated antigen 520 homolog
Gene Symbols CHP2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 146-191 of Human CHP2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308965, 63928, 70261
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.