Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRNA1 Monoclonal Antibody (88B)
GREENER_CHOICE

Catalog No. MA3043 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA3043 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA3043 Supplier Invitrogen™ Supplier No. MA3043
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA3-043 detects nicotinic acetylcholine receptor (nAChR) gamma and delta subunits in torpedo and the delta subunit in mouse, human, rat, chicken and frog. This antibody does not detect the alpha 1 and beta 1 subunits. MA3-043 has been successfully used in Western blot, immunohistochemistry and immunoprecipitation procedures. By Western blot, this antibody detects a 60 kDa protein representing the gamma subunit and a 65 kDa protein representing the delta subunit from torpedo skeletal muscle homogenates. Under non-reducing conditions the delta subunit migrates mostly as a dimer of ~130 kDa. Immunohistochemical staining of nAChR in rat skeletal muscle with MA3-043 results in strong staining of the motor endplate. In Immunohistochemical staining, cryostat or permeabilization of the sections is recommended due to the reactivity of this antibody with the cytoplasmic side of the receptor. The MA3-043 antigen is purified Torpedo californica acetylcholine receptor.

Acetylcholine receptors (AChRs) mediate synaptic transmission at the neuromuscular junction. The channel-linked AChR that mediates rapid, excitatory actions of acetylcholine is called nicotinic AChR (nAChR) because it can be activated by nicotine. The non-channel linked AChR that medicates the slow actions of acetylcholine, which can be either inhibitory or excitatory, is called muscarinic AChR (mAChR) because it can be activated by muscarine.The nAChRs fall into two subclasses, the muscle-type and the neuronal-type. The muscle-type receptor is a pentameric glycoprotein of five membrane spanning subunits, (2 alpha 1's, beta 1, gamma or epsilon and delta) which form a ligand gated ion channel. The neuronal nAChR is also a pentamer, but unlike the muscle-type, is made up of only two general types of subunits, alpha and beta of which multiple subtypes have been described (alpha 2 to alpha 8 and beta 2 to beta 4).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNA1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 88B
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.05% sodium azide
Gene CHRNA1
Gene Accession No. P02708, P04756, P09479, P25108
Gene Alias acetylcholine receptor; Acetylcholine receptor subunit alpha; acetylcholine receptor, nicotinic, alpha 1 (muscle); Achr-1; ACHRA; ACHRD; Acra; AI385656; AI608266; alpha-1 subunit, nicotinic acetylcholine receptor; CHNRA; cholinergic receptor; cholinergic receptor nicotinic alpha 1 subunit; cholinergic receptor nicotinic alpha polypeptide 1; cholinergic receptor, nicotinic alpha 1; cholinergic receptor, nicotinic, alpha 1 (muscle); cholinergic receptor, nicotinic, alpha polypeptide 1 (muscle); CHRNA; CHRNA1; CMS1A; CMS1B; CMS2A; FCCMS; muscle nicotinic acetylcholine receptor; nAChR; NACHRA1; nicotinic acetylcholine receptor alpha subunit; nicotinic acetylcholine receptor, alpha-1 subunit; nicotinic cholinergic receptor alpha 1; SCCMS
Gene Symbols CHRNA1
Host Species Mouse
Immunogen Purified Torpedo californica acetylcholine receptor.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1134, 11435, 395608, 79557
Target Species Human, Mouse, Rat, Chicken, Amphibian, Fish
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.