Learn More
Clontech Labs 3P CHROMA SPIN+TE-1000 Columns, 50 each
SDP
Supplier:  Clontech Labs 3P 636079
Encompass
CHROMA SPIN+TE-1000 Columns are used for the purification of nucleic acids larger than 1,000 bp in size. Matrix is in TE buffer (10 mM Tris-HCl (pH 8.0), and 1 mM EDTA).
Catalog No. NC1753167
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.