Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Chromacol Vial Racks

Catalog No. 13622423
Change view
Click to view available options
Holds:
Vials
Vials (100)
Material:
Aluminum
Plastic
For Use With (Equipment):
12 mm OD Vials
15 mm OD Vials
22-23 mm OD Vials
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Material For Use With (Equipment)
13-622-423 Vials Aluminum 22-23 mm OD Vials
13-622-404 Vials (100) Plastic 12 mm OD Vials
13-622-424 Vials Aluminum 15 mm OD Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-622-423 Supplier Thermo Scientific™ Supplier No. T28

Ensure a safe working position on the lab bench and during transport with these robust vial racks.

Save space in the fridge while ensuring a safe working position on the bench and during transport. Available in a various materials, colors and capacities, Thermo Scientific™ Chromacol Vial Racks meet a variety of application needs

Choose from an assortment of chemically resistant and fairly robust racks.

  • Aluminum and plastic models are temperature resistant from -90 to +121°C (-130 to +250°F)
  • Aluminum and plastic models are autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Description Vial Rack
Autoclavable Autoclavable
Color Silver
Holds Vials
Material Aluminum
Positions 28
For Use With (Equipment) 22-23 mm OD Vials
Product Type Vial Rack

Limited warranty period:
Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.