Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ CHROMAFLEX™ 1.0 cm Jacket Sealing Ring
SDP

Catalog No. 12141405 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With:
420870-1000, 420870-1200, 420870-1500, 420870-3000, 420780-6000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
12-141-405 420870-1000, 420870-1200, 420870-1500, 420870-3000, 420780-6000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-141-405 Supplier DWK Life Sciences Supplier No. 4208381000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replacement Parts - Jacket Sealing Ring for 420870 series 1.0 cm columns

1.0 cm Jacket Sealing Ring for 420870-1000, 420870-1200, 420870-1500, 420870-3000, 420870-6000
TRUSTED_SUSTAINABILITY

Specifications

For Use With 420870-1000, 420870-1200, 420870-1500, 420870-3000, 420780-6000
Product Type Jacket Sealing Ring
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.