Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences KIMBLE™ CHROMAFLEX™ Flow Adapters
SDP

Catalog No. K4208761620 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric) Inner:
1 cm
2.5 cm
4.8 cm
Diameter (English) Outer:
0.063 in.
0.125 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner Diameter (English) Outer
K4208761620 2.5 cm 0.063 in.
K4208760000 1 cm 0.063 in.
K4208760020 2.5 cm 0.125 in.
K4208760040 4.8 cm 0.125 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. K4208761620 Supplier DWK Life Sciences Supplier No. 4208761620
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KIMBLE™ CHROMAFLEX™ Flow Adapters improve chromatogram resolution by delivering sample and buffer directly to the gel bed top, protecting it from disruption during loading and eliminating dead volume above the gel bed.

  • Flow adapters are recommended when using buffer gradients, during reverse flow applications and for high resolution gravity flow separations
  • Each flow adapter is supplied with a 20 micron porosity HDPE bed support, TFE/propylene o-ring seal, stainless steel adjusting rod, PTFE body, and 5 feet (1.5 meters) of FEP PTFE tubing
  • Blue lock ring is supplied with all sizes except those that fit 1.0 cm column ID
  • Use of aggressive organic solvents requires the PTFE bed supports listed as accessories and FFKM o-rings sizes 010, 116 or 222
TRUSTED_SUSTAINABILITY

Specifications

Type Adapter
Diameter (English) Outer 0.063 in.
Diameter (Metric) Inner 2.5 cm
Temperature (English) Max. 122°F
For Use With (Application) Improving Chromatogram Resolution
Includes Blue lock ring, 20 micron porosity HDPE bed support, TFE/propylene o-ring seal, stainless steel adjusting rod, PTFE body, and 5 ft. (1.5 meters) of FEP PTFE tubing
Description Fits column ID: 2.5 cm
For Use With Column, Acrylic Jacketed
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.