Learn More
Liofilchem™ Chromatic MH
SDP
Catalog No. 21403473 Shop All Liofilchem Products
Change view
Click to view available options
Quantity:
20 Plates
Catalog No. Quantity
21-403-473 20 Plates
1 options
Catalog No. 21-403-473 Supplier Liofilchem™ Supplier No. 11618
Add to Cart
Add to Cart
Mueller Hinton agar is a medium for the microbial differentiation simultaneously with susceptibility testing directly from clinical specimens
  • Chromogenic Muller Hinton for presumptive identification and susceptibility testing of various microorganisms from clinical specimens.
  • In the Intensive Care Unit the mortality rates for VAP, sepsis, surgical site or intra-abdominal, catheter related infections are critically high. Direct M.I.C. on CSF, positive blood culture bottles and other specimens from critical patients and direct M.I.C. on bronchial aspirates from patients with VAP can contribute with timely and essential information to save the life of patients.
Specifications
Manufacturing Origin Italy
Product Type Prepared Microbiology Plate
Sterility Non-sterile
Target Organism Class Bacteria, Gram-negative Organisms, Gram-positive Organisms
Test Type Microbial Differentiation, Antimicrobial Susceptibility
Quantity 20 Plates
Diameter (Metric) 90 mm