Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chromatin Immunoprecipitation (ChIP) Assay Kit
GSA_VA

Catalog No. 17295 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
22 Assays/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-295 22 Assays/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-295 Supplier MilliporeSigma™ Supplier No. 17295
Add to Cart
Add to Cart

Includes

  • Protein A agarose/Salmon Sperm DNA
  • All necessary buffers

Used to enrich for specific regions of chromatin that are immunoprecipitated with a specific antibody for a chromatin protein. Works with all eukaryotic cell types

  • Contains all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions 
  • Supplied buffers are sufficient to generate chromatin from up to five 15 cm plates of cultured cells, each plate providing up to 10 chromatin preparations (varies with cell and assay type)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) 22 individual chromatin immunoprecipitation (ChIP) reactions using inexpensive protein A agarose beads
Quantity 22 Assays/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.