Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Chromatography Strips
SDPSCI_ED

Catalog No. s23973 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S23973 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S23973 Supplier United Scientific™ Supplier No. CHRP100
Only null left
Add to Cart
Edge
Add to Cart

Used in chromatography studies. These Chromatography Strips are 15 x 2 cm in size.

  • Available in size L x W: 15 x 2 cm.
  • Sold as a pack of 100.

Specifications

Grade 5+
For Use With (Application) Chromatography Studies
Includes A pack of 100 15 cm long by 2 cm wide strips used in chromatography studies.
Quantity 100 Pk.
Class Size 10 Lab Groups
Product Type Chromatography Strips
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.