Learn More
MP Biomedicals™ ChromatoPur™ Bovine Thrombin, Low Specific Activity, Diagnostic Grade
Shop All MP Biomedicals, Inc ProductsDescription
Chromatographically purified with minimal loss of native clotting activity and minimized degradation to the gamma form. Enzyme activity is measured by clotting assay. 90 to 300U/mg protein.
Physical Characteristics
- Appearance: White to off-white powder
- pH: 7.0 to 8.0
- Protein: ≥50%
- Specific activity: 90 to 300U/mg protein
- Activity: Report μ/mg solid
- Total Activity: U/vial
- Exipients: Reported
Specifications
Specifications
| For Use With (Application) | Cell Culture, Microbial Culture |
| Name | Bovine Thrombin |
| pH Range | 7 to 8 |
| Quantity | 25 kU |
| Source | Bovine Thrombin |
| Storage Requirements | -10°C |
| Form | Solid |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.