Learn More
HexArmor™ Chrome Series™ 4080
Catalog No. 19162070 Shop All Performance Fabrics, Inc Products
Click to view available options
Size:
2X-Large
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-162-070 2X-Large
19-162-071 Large
19-162-068 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-162-070 Supplier HexArmor™ Supplier No. 4080XXL 11
Add to Cart
Add to Cart
This item is not returnable. View return policy
Leather impact gloves combine comfort with modern protection using an interior layer of SuperFabric™ brand material and an abrasion resistant leather shell, providing hands with high levels of cut and puncture protection.
  • SuperFabric™ brand material palm provides ANSI/ISEA level A8 cut resistance for some of the highest cut-resistance available in the industry (interior layer)
  • Hi-vis back-of-hand impact protection system utilizes an advanced design to dissipate forceful blows over a large area
  • EN407 level 3 contact heat rated  Goatskin leather provides a traditional style of comfort and grip
  • Velcro™ cuff for a comfortable and precise fit
Specifications
Certifications/Compliance ANSI/ISEA
Color Gray, Fluorescent Yellow
Cut Resistance Rating ANSI A8
Material Leather, SuperFabric
Product Type Cut-resistant Gloves
Size 2X-Large
EN388 Standard EN 388:4X13FP
Glove Cuff Style Hook and Loop
Model 4080-XXL
Series Chrome Series™ 4080
Length (English) 9.8 in.
Length (Metric) 25 cm