Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromic Acid, 30% w/v With Iron, in 0.4% v/v Sulfuric Acid, Reagents

Catalog No. 01335643
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-643 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-643 Supplier Reagents Holdings Llc Supplier No. CC120300500A
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chromic Acid, 30% w/v With Iron, in 0.4% v/v Sulfuric Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1333-82-0 , 1002577-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Chromium trioxide solution
IUPAC Name water
SMILES O

Specifications

pH <1
Color Red
Boiling Point 106°C
Packaging Poly Bottle
Specific Gravity 1.1908
Quantity 500 mL
Physical Form Liquid
Percent Purity 30%
Grade Reagent
Chemical Name or Material Chromic Acid, 30% w/v With Iron, in 0.4% v/v Sulfuric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.