Learn More
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Color | Red |
| CAS Max % | 100 |
| Melting Point | 196°C |
| Density | 2,700kg/m3 |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Specific Gravity | 2700kg/m3 |
| Linear Formula | CrO3 |
| Quantity | 500 g |
| UN Number | UN1463 |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement May cause fire or explosion; strong oxidizer. Fatal if inhaled. Toxic if swallowed or in contact with skin. Causes severe skin burns and eye damage. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. May cause genetic defects. May cause cancer. Suspected of damaging fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Do not handle until all safety precautions have been read and understood. Keep away from heat, sparks, open flames. No smoking. Keep away from clothing, combustible materials. Take any precaution to avoid mixing with combustibles. Do not breathe dust. Wash exposed skin thoroughly after handling. Do not eat, drink or smoke when using this product. Use only outdoors or in a well-ventilated area. Contaminated work clothing must not be allowed out of the workplace. Avoid release to the environment. Wear protective gloves, flame resistant protective clothing, eye protection, face protection. Wear respiratory protection. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/doctor. If on skin, hair or clothing: Rinse immediately contaminated clothing and skin with plenty of water before removing clothes. Rinse skin with water/shower. Immediately call a poison center/doctor. If inhaled: Remove person to fresh air and keep comfortable for breathing. Immediately call a poison center/doctor. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. In case of fire: Use carbon dioxide, powder, alcohol-resistant foam to extinguish. In case of major fire and large quantities: Evacuate area. Fight fire remotely due to the risk of explosion. Collect spillage. Store locked up in a well-ventilated place. Keep container tightly closed. Dispose of contents/container to comply with local, state and federal regulations. |
| EINECSNumber | 215-607-8 |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
