Learn More
SPEX CERTIPREP LLC 3P CHROMIUM 2% HNO3 1000PPM 100ML
Supplier:  SPEX CERTIPREP LLC 3P 1000121100
Encompass_Preferred
(HPS by Zeptometrix) Chromium, Appearance/Odor: A lustrous, hard, brittle, metallic element, Boiling Point: 100degreeC, Specific Gravity: 1.0020, Solubility in Water: Complete, Keep container tightly closed
Catalog No. NC9139852