Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium boride, 99% (metals basis)

Catalog No. AA1257014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1257014 25 g
AA1257022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1257014 Supplier Thermo Scientific Chemicals Supplier No. 01257014
Add to Cart

CAS: 12007-16-8 | CrB2

Chromium boride,is been used in plasma technology and for high-performance electrodes in electron microscopes, metal matrix composites (MMCs), cutting tools and cermets for machining aluminum. chromium diboride is recognised as offering superior performance in corrosive-wear environments. Chromium diboride thin films possess desirable combinations of properties, which are attractive for a wide range of potential industrial applications. As a wear-resistant component of weld materials and powders for spraying in engineering and metallurgy, Protection of outer surfaces and bearing surfaces of machine parts against abrasive wear by plasma spraying, In aluminum evaporators manufacturing and for aluminum electrolyzes repair work, In nuclear engineering as neutron absorption material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12007-16-8
Molecular Formula CrB2
MDL Number MFCD00016043

Specifications

Color Gray
Physical Form Powder
Quantity 25 g
Assay Percent Range 99% (Metals basis)
Solubility Information It is insoluble in water.
Formula Weight 73.62
Odor Odorless
Chemical Name or Material Chromium boride
Hazard Category H317-H350i
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 234-499-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.