Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(III) 2-ethylhexanoate, 50% in 2-ethylhexanoic acid

Catalog No. AAA1656018
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1656018 50 g
AAA1656030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1656018 Supplier Thermo Scientific Chemicals Supplier No. A1656018
Add to Cart

CAS: 3444-17-5 | C24H45CrO6 | 481.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3444-17-5
Molecular Formula C24H45CrO6
Molecular Weight (g/mol) 481.61
MDL Number MFCD00014005
InChI Key WBKDDMYJLXVBNI-UHFFFAOYNA-K
Synonym chromium iii 2-ethylhexanoate, chromium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, chromium 3+ salt 3:1, chromic 2-ethylhexanoate, chromium tris 2-ethylhexanoate, chromium 2-ethylhexanoate 1:3, 2-ethylhexanoic acid chromium iii salt, hexanoic acid, 2-ethyl-, chromium 3+ salt, chromium 3+ ; 2-ethylhexanoate
PubChem CID 103012
IUPAC Name chromium(3+);2-ethylhexanoate
SMILES [Cr+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O

Specifications

Odor Weak, Characteristic
Quantity 50 g
Formula Weight 481.62
Concentration or Composition (by Analyte or Components) 50% in 2-ethylhexanoic acid
Chemical Name or Material Chromium(III) 2-ethylhexanoate
Hazard Category H361d
Hazard Statement GHS H Statement
H361-H315-H319
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 222-357-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.