Learn More
Chromium(III) oxide, Puratronic™, 99.97% (metals basis)
Description
Also known as green compound for buffing and polishing, it can be used as abrasives. Apart from being used as a precursor to the magnetic pigment viz., chromium dioxide, it is used in inks, glasses, and pigments (viridian). They are employed as plating agents and surface treating agents. It is a catalyst in the Phillips process of manufacture of high density polythene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 3000°C |
| Melting Point | 2435°C |
| Color | Green |
| Physical Form | Powder |
| Quantity | 25 g |
| Merck Index | 14,2234 |
| Solubility Information | Insoluble in water,alcohol,acetone,alkalies and acids. |
| IUPAC Name | dichromium(3+) trioxidandiide |
| Formula Weight | 151.99 |
| Percent Purity | 99.97% |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H302-H332-H317 Harmful if swallowed. Harmful if inhaled. May cause an allergic skin reaction. |
| Precautionary Statement | GHS P Statement P261-P280-P304+P340-P301+P312-P312-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Call a POISON CENTER or doctor/physician if you feel unwell. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 215-160-9 |
| RTECSNumber | GB6475000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.