Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium Potassium Sulfate, Dodecahydrate, Crystal, ACS, 98-102%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
7788-99-0
7788-99-0
Grade ACS
CAS Max % 1
Aluminum (Al) 0.0002
Ammonium 0.0001
Percent Purity 98 to 102%
Chloride 2e-05
Heavy Metals 0.0001
Insoluble Matter 0.0001
Iron (Fe) 0.0001
Packaging Fiber Drum
Quantity 50 kg
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.