Learn More
Chromium Trioxide, ACS Reagent (Chromic Acid), Reagents
Specifications
Specifications
| Boiling Point | 250°C |
| Melting Point | 196°C |
| Color | Dark red-Black |
| Physical Form | Solid |
| Quantity | 500 g |
| Grade | ACS |
| Packaging | Glass Bottle |
| Specific Gravity | 2.7 |
| Chemical Name or Material | Chromium Trioxide, ACS Reagent (Chromic Acid) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.