Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium Trioxide, Technical Flake (Chromic Acid), Reagents

Catalog No. 01337316
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-316 2.5 kg
01-337-317 100 g
01-337-314 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-316 Supplier Reagents Holdings Llc Supplier No. C21795002.5KG1
Add to Cart
Add to Cart

Chromium Trioxide, Technical Flake (Chromic Acid), Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1333-82-0
Molecular Formula CrO3
Molecular Weight (g/mol) 99.99
MDL Number MFCD00010952
InChI Key WGLPBDUCMAPZCE-UHFFFAOYSA-N
Synonym Chromic Acid
IUPAC Name trioxochromium
SMILES O=[Cr](=O)=O

Specifications

Boiling Point 250°C
Melting Point 196°C
Color Dark Red-Black
Physical Form Solid
Quantity 2.5 kg
Grade Technical
Packaging Glass Bottle
Specific Gravity 2.7
Chemical Name or Material Chromium Trioxide, Technical Flake (Chromic Acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.