Learn More
Bachem H-D-Phe-Homopro-Arg-pNA · 2 acetate
SDP
Supplier:  Bachem 4008145.0250
Encompass_Preferred
H-D-Phe-Homopro-Arg-pNA · 2 acetate, 386253-60-7, C27H36N8O5 · 2 C2H4O2, Mr 672.74, 250mg. H-D-Phe-Pip-Arg-pNA · 2 acetate, H-D-Phe-L-pipecolyl-Arg-pNA · 2 acetate. Chromogenic substrate specific for thrombin but insensitive to factor Xa. Kinetic data for Thrombin: kcat= 91.4, Km = 1.33 uM.
Catalog No. 50-194-6936
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.