Learn More
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Monkey, Pig, Rat and Rabbit. Others not known
Epitope: Not determined
Immunogen: Recombinant protein encoding human chromogranin A
Molecular Weight: 68-75 kDa
Positive Control: Pancreas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Chromogranin A |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Clone | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encoding human chromogranin A |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Research Discipline | Oncogenesis |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.