Learn More
MilliporeSigma™ Chromolith™ DIOL5 Guard Cartridges, 5-4.6
Catalog No. M1531750001 Shop All MilliporeSigma Products
Change view
Click to view available options
DIOL5 phase
Catalog No.
M1531750001 DIOL5 phase
1 options
Catalog No. M1531750001 Supplier MilliporeSigma™ Supplier No. 1.53175.0001
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Functions in reverse phase (when in RP mode) or normal phase. MilliporeSigma™ Chromolith™ DIOL5 Guard Cartridges, 5-4.6 come in 3 pieces with 5mmL x 4.6mmID.
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 5 mm
Diameter (Metric) Inner 4.6 mm
Stationary Phase DIOL5
Phase RP (reversed-phase, when in mode) & NP (normal-phase)
Column Type Chromolith™ HPLC guard cartridges