Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Chromotrope 2B

Catalog No. AAL1133022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1133022 100 g
AAL1133014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1133022 Supplier Thermo Scientific Chemicals Supplier No. L1133022
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 548-80-1 | C16H9N3Na2O10S2 | 513.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 548-80-1
Molecular Formula C16H9N3Na2O10S2
Molecular Weight (g/mol) 513.36
MDL Number MFCD00003951
InChI Key MQHFURHHKNKIRN-GVLVODDDSA-L
Synonym Acid Red 176; C.I. 16575
PubChem CID 44135472
SMILES [Na+].[Na+].OC1=CC(=CC2=C1C(=O)\C(=N/NC1=CC=C(C=C1)[N+]([O-])=O)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Melting Point >300°C
Beilstein 4109733
Merck Index 14,2240
Quantity 100 g
IUPAC Name disodium (3E)-5-hydroxy-3-[2-(4-nitrophenyl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Formula Weight 513.37
Percent Purity 97%
Chemical Name or Material Chromotrope 2B
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-959-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.