Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Chromotrope 2B, pure

Catalog No. AC149750050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC149750050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC149750050 Supplier Thermo Scientific Chemicals Supplier No. 149750050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 548-80-1 | C16H9N3Na2O10S2 | 513.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 548-80-1
Molecular Formula C16H9N3Na2O10S2
Molecular Weight (g/mol) 513.36
MDL Number MFCD00003951
InChI Key MQHFURHHKNKIRN-GVLVODDDSA-L
Synonym Acid Red 176, C.I. 16575, 4,5-Dihydroxy-3-(p-nitrophenylazo)-2,7-naphthalenedisulfonic acid, disodium salt
PubChem CID 44135472
IUPAC Name disodium (3E)-5-hydroxy-3-[2-(4-nitrophenyl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
SMILES [Na+].[Na+].OC1=CC(=CC2=C1C(=O)\C(=N/NC1=CC=C(C=C1)[N+]([O-])=O)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Melting Point 300.0°C
Infrared Spectrum Authentic
Lambda Max 513 to 517nm (c=0.1, H2O)
Loss on Drying 18% max.
Quantity 5 g
Formula Weight 513.35
Grade Pure
Chemical Name or Material Chromotrope 2B
Hazard Category Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.