Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromotropic Acid Disodium Salt, ACS, Spectrum™ Chemical
SDP

Catalog No. 18609797 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-797 100 g Amber Glass Bottle
18-609-795 5 g Amber Glass Bottle
18-609-796 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-609-797 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1629100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5808-22-0

Specifications

Physical Form Crystalline Powder
Quantity 100 g
Molecular Weight (g/mol) 400.29
Grade ACS
Packaging Amber Glass Bottle
Chemical Name or Material Chromotropic Acid Disodium Salt

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.