Learn More
Chromplete™ Acetonitrile, HPLC/GC, meets ACS/USP analytical requirements
Shop All Fisherbrand ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chromplete Acetonitrile meets the analytical requirements of USP and is suitable for use in:
- HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
- GC- FID, GC- ECD, GC-TCD
- Pesticide Analysis by GC or HPLC
- UV Spectrophotometry
- Preparative chromatography
- Supercritical Fluid and Thin Layer chromatography
- Reactions/Analyses requiring ACS compliance
- Extraction/Concentration Analyses
Specifications
Specifications
| Melting Point | -51°F |
| Absorbance | <0.1 at 195 nm, <0.05 at 200 nm, <0.02 at 210 nm, <0.01 at 220 nm |
| Color | ≤ 5 APHA |
| CAS Min % | 0.9995 |
| Density | 0.775 - 0.78 |
| Quantity | 4 L |
| Packaging | Aluminum Jug |
| Physical Form | Liquid |
| Refractive Index | 1.3405 – 1.3425 |
| Residue after Evaporation | ≤ 0.8 ppm |
| Show More |
Safety and Handling
| Hazard Category | Serious eye damage/eye irritation (category 2), Flammable liquids (category 2), Acute toxicity (category 4) |
| Hazard Statement | Causes serious eye irritation, Highly flammable liquid and vapor, Harmful if swallowed, in contact with skin or if inhaled |
| ShelfLife | 1825 days |
| EINECSNumber | 200-835-2 |
| RTECSNumber | AL7700000 |
| storageNote1 | In sealed containers |
| Recommended Storage | Room temperature flammable storage |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.