Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromplete™ Methanol, HPLC/GC, meets ACS/USP analytical requirements

Catalog No. t001021000 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T001021000 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T001021000

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Methanol is suitable for HPLC, GC, SFC, TLC, ACS, and Spectrophotometry. | CAS: 67-56-1 | CH4O | 32.04 g/mol

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Methanol is suitable for HPLC, GC, SFC, TLC, ACS, and Spectrophotometry.

Chromplete Methanol meets the analytical requirements of USP and is suitable for use in:

  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • Pesticide Analysis by GC or HPLC
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Absorbance ≤1.00, ≤ 0.23, ≤ 0.11, ≤ 0.01
Color ≤ 5 APHA
CAS Min % 0.999
Density 0.775 – 0.780
Quantity 1 L
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation ≤ 1.0 ppm
Titratable Acid ≤0.0003 meq/g
Titratable Base ≤0.0002 meq/g
UN Number 1230
Water ≤0.07%
Boiling Point 148.5°F
Formula Weight 32.04
Percent Purity 99.9%
Grade Chromplete
Assay Minimum, GC-FID
Chemical Name or Material Methanol
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Flammable liquids (category 2), Specific target organ toxicity after single exposure (category 1), Acute toxicity (category 3)
Hazard Statement Highly flammable liquid and vapor, Causes damage to organs, Toxic if swallowed, in contact with skin or if inhaled
ShelfLife 1825 days
EINECSNumber 200-659-6
RTECSNumber PC1400000
storageNote1 In sealed containers
Recommended Storage Room temperature flammable storage

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.