Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHST13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140122
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140122 Supplier Invitrogen™ Supplier No. PA5140122
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GNRALGSSWL GGEKRSPLQK LYDLDQDPRS TLAKVHRQRR DLLNSACSRH SRRQRLLQPE DLRHVLVDDA HGLLYCY Highest antigen sequence indentity to the following orthologs: Mouse - 77%, Rat - 73%.

The protein encoded by this gene belongs to the sulfotransferase 2 family. It is localized to the Golgi membrane, and catalyzes the transfer of sulfate to the C4 hydroxyl of beta-1, 4-linked N-acetylgalactosamine (GalNAc) flanked by glucuronic acid residue in chondroitin. Chondroitin sulfate constitutes the predominant proteoglycan present in cartilage and is distributed on the surfaces of many cells and extracellular matrices.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST13
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHST13
Gene Accession No. Q8NET6
Gene Alias 1110067M19Rik; C4ST3; C4ST-3; carbohydrate (chondroitin 4) sulfotransferase 13; carbohydrate sulfotransferase 13; Chondroitin 4-O-sulfotransferase 3; chondroitin 4-sulfotransferase 3; CHST13; DcR3; Decoy receptor 3; M68; RGD1562825
Gene Symbols CHST13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHST13. Recombinant protein control fragment (Product #RP-109145).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166012
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.