Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHST2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735172 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5172 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5172 Supplier Proteintech Group Inc Supplier No. 260271AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody recognizes both short form and long form, which migrates around 50-52 kDa and 58-60 kDa, respectively.

N-acetylglucosamine-6-O-sulfotransferases, such as CHST2, catalyze the transfer of sulfate from 3-prime-phosphoadenosine 5-prime-phosphosulfate.

Specifications

Antigen CHST2
Applications Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CHST2
Gene Accession No. Q80WV3, Q9Y4C5
Gene Alias C6ST, CHST2, GlcNAc6ST 1, GN6ST, Gn6ST 1, GST 2, GST2
Gene Symbols Chst2
Host Species Rabbit
Immunogen CHST2 Fusion Protein Ag23293
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54371, 9435
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.